Ward's® Zygospore Formation Sets
Supplier: VWR
470179-620EA 29.95 USD
470179-620 470179-618
Ward's® Zygospore Formation Sets
Specimens Live Specimens Fungi - Living Material
Observe Results of Fungal Sexual Reproduction
Your choice of two sexually reproducing fungi are available in demonstration plates; each set contains a “+” strain on one half of the plate and a “-” strain on the other half, with a line of zygospores already formed at the junction of the mycelia. Each set comes with a teacher’s guide and the Working with Fungi manual. Note: Allow two weeks for delivery.