Ward's® Live Zebrafish
Supplier: VWR
Ward's®
WARD470324-864 WARD470176-328 WARD470324-866 WARD470180-560 WARD470180-552 WARD470324-874 WARD470324-872 WARD470324-870 WARD470324-868
470180-552KT 59.95 USD
470180-552 470324-870 470324-872 470324-864 470324-874 470324-866 470324-868 470176-328 470180-560
Ward's® Live Zebrafish
Specimens Live Specimens Aquaria - Living Material
Brachydanio rerio.
These striped fish are prolific breeders, laying transparent eggs that are ideal for observing development. Size: approximately 1".
Packaging: Gender specific packages available.