Would you like to log in again?
Young's Double Slit Apparatus
Supplier: SCIENCE FIRST INC SE
22300
Large scale model of Young's double slit experiment.
  • 20,000 times larger than the light phenomena
  • Easy to see wavelengths
Built 20,000 time larger that the actual experimental conditions of a 600 nm HeNe laser illuminating a slit of 4.5 or 2.25 microns, this apparatus allows calculations to be made for this classical investigation.
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Young's Double Slit Apparatus 30-1000272-00 470006-476 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Dimensions 57 x 40 cm
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty