Working Nephron Model
WARD470101-636
470101-636KT 169.99 USD
470101-636
Working Nephron Model
Developed by the Bert half of the Wartski team, this wet lab effectively demonstrates active transport, dialysis, osmosis, diffusion, filtering, and the many forces of kidneys.
Students observe how a nephron functions and test simulated blood, urine and body solution for pH, glucose, protein, and starch. Includes instructions. Grades 10–12.