Would you like to log in again?
Weather In a Beaker: Relative Humidity & the Orographic Effect
Supplier: VWR
Visually Capture the Workings of the Orographic Effect
A series of beakers set on ring stands set against a chart depicting leeward and wayward sides of a mountain and the Orogrphic Effect helps students understand relative humidity as it pertains to air temperature. Water capacity and volume of air increases with an increase in air temperature, however, air density decreases with air temperature. As the air is forced upward over a mountain range, it cools, causing water vapor to condense and rain out. As the air flows down the other side of the mountain, it warms, promoting evaporation and creating a desert climate. Developed by science teacher, Tom Hoolihan, this simple demonstration is extremely effective in demonstrating a complex concept. Includes instructions.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Weather in a Beaker: Relative Humidity & the Orographic Effect   470004-556 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Base Dimensions Rod Length Description Overall Dimensions Rod O.D. Supplier No. VWR Catalog Number Unit Price Quantity
- 4 x 6" - 18" SSB4X6CI SSB4X6CI 470019-526 Each Retrieving
Plain - 5" x 5" - WGPL55-P12 WGPL55-P12 470019-500 Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty