Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Ward's® Volumeter
Supplier: VWR
Measure photosynthetic rate or respiration.
Detect small changes in volume resulting from the exchange of gases in photosynthesis or respiration. Two volumeters and a thermobarometer are suspended in a single vessel, which serves as a one-gallon waterbath. Place the living specimen in one or both of the clear volumeter tubes and a drop of colored water introduced into the system moves along a calibrated tube in response to oxygen/carbon dioxide exchange of the specimen. Instructions included.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
WARD’S Volumeter   470019-318 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty