Would you like to log in again?
Vital Stain Set 3
Supplier: VWR
Intracellular Structures Stained for Easy Viewing
Vital staining highlights individual cell structures without killing the cell, making cell morphology easier to understand. Our unique polychrome method (ProtoColor) allows cytoplasmic structures to be clearly observed using brightfield microscopy. Set comes with the Working with Protozoa manual and materials for 35 students.
Cultures: Amoeba, Bursaria truncatella, Chaos or Pelomyxa, Hydra, Paramecium caudatum, Stentor, and Vorticella.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Vital Stain Set 3   470180-222 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty