Vital-stained Live Specimen Sets
Supplier: Avantor
WARD470180-220 WARD470180-218
470180-218KT 27.95 USD
470180-218 470180-220
Vital-stained Live Specimen Sets
Specimens Live Specimens Protists - Living Material
Sets of aquatic specimens, stained for easy viewing
  • Individual cell structures are easier to see than in unstained cultures
  • Staining is harmless to the organism
  • ProtoColor lets cytoplasmic structures be easily observed using brightfield microscopy
  • Choose from multiple set options.
  • Many other vital stained specimens available separately
Set 1 contains the following 3 cultures: Amoeba proteus, Paramecium caudatum, Hydra littoralis
Set 2 contains the following 5 cultures: Amoeba proteus, Paramecium caudatum, Bursaria truncatella, Stentor sp., Vorticella sp.