Would you like to log in again?
Vacuum Plate for Bell Jars
Create a vacuum with the tight-sealing, corrosion-resistant polyethylene plate with rubber mat. A fitting on the side accommodates hoses with 1/4"inner dia. Size 10 1/2" square, 3/4" thick.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Vacuum Plate for Bell Jars 7-203-7 470020-808 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty