Utility Tongs
3511
470018-926EA 3.85 USD
470018-926
Utility Tongs
These tongs are designed for handling cylindrical apparatus such as test tubes, flasks, and small beakers up to 2" in diameter.
  • Clamping edges of the jaws are flattened with serrations to prevent slipping
  • Constructed of nickel-plated steel
  • Size: 8"L