Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® TI Light Probe

Supplier: VERNIER SOFTWARE & TECH SE
TILT-BTA
470017-606
Vernier® TI Light Probe
Best for Comparative or Relative Measurements.

  • Measure light
  • Investigate inverse square law of light

This light sensor uses a phototransistor to measure irradiance and UV, visible and infrared light. Complete investigations outside the visible light spectrum with the infrared capabilities of the sensor. This probe provides enables you to investigate the inverse square law of light, measure changes in light intensity for crossed polarizers and much more.

Specifications:

Range: 10 μW/cm2 to 1 mW/cm2

Accuracy: ±20%

Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty