The Brain Microslide
T-237
470100-848EA 10 USD
470100-848
The Brain Microslide
Investigate the brain and its related structures
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson plan on the topic under study
This set reveals the structure of the brain including MRI "slices" of the author's brain and detailed images made of slides of brain tissues.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Content:
The Whole Brain (MRI)
Cerebrum (MRI)/Pyramidal Cells (250x)
Axions in the Corpus Callosum (400x)
Thalamus (MRI)/Nerve Tracts (750x)
Eyes & Occipital Lobes (MRI)/Optic Nerve (400x)
Motor & Sensory Areas (MRI)/ Motor Neurons in Brain (250x)
“Crossover of Nerves” (400x)
Purkinje Cells (100x)