Preserved Squid
470158-646EA 60 USD
470158-646 470158-644 470158-642 470158-640
Preserved Squid
Specimens Preserved Specimens Display Specimens
Formalin-preserved squid are available in pails.
  • Mollusc Dissection
  • Advanced Nervous System
  • For Eye Anatomy
  • Formalin-Preserved
  • Dissection Shows Nervous System
The squid,LoligoorOmmastrephes, has the most highly developed nervous system of all the invertebrates.
Dissection shows the developed eyes and nervous system.