Disposable Cardboard Mouthpiece Spirometer, Propper Manufacturing
242001
470206-378CS 65.8 USD
470206-378
Disposable Cardboard Mouthpiece Spirometer, Propper Manufacturing
Propper single use disposable spirometer mouthpieces are an economical option to help reduce risk of contamination.
  • Single-use disposable cardboard mouthpieces
  • Significantly reduces the risk of cross-infection between patients
Simply blow into the spirometer and read vital lung capacity measurement on the dial. The device is useful for comparing volumes between students and calculating average volume and variance, or for comparing volumes before and after exercise. It comes with 50 disposable mouthpiece tubes and instructions that include vital capacity charts.
Range: 0–7 L. Size: 2 1/2 in. dia. x 3 1/4 in.H.
Single use disposable spirometer mouthpieces. Disposable nature assists in the reduction of cross contamination between patients. Compatible with most spirometers. The internal diameter and external diameter is 9 mm and 12 mm.
Ordering information: Note: these spirometer mouthpieces are not compatible with the 9L Student Wet Spirometer (470007-990).