Ward's® Simulated Red Blood Cell Counts With a Hemacytometer
Supplier: Ward's
WARD470023-672
470023-672EA 48.95 USD
470023-672
Ward's® Simulated Red Blood Cell Counts With a Hemacytometer
Educational Materials Biology Educational Materials Blood Typing Learning Activities
Diagnose Anemia from Unknown Samples
Students gain experience in taking red blood cell counts safely with WARD’S Simulated Blood. Materials for up to 2,000 counts and instructions are included. A hemocytometer, required, is available separately.
Accessories information: This simulated blood will need to be diluted, just like real blood, for a proper count. If you do not already have dilution pipets, please consider our Blood Cells Diluting Pipets