Ward's® Serpentinite
Supplier: VWR
WARD470026-132 WARD470026-130 WARD470026-128
470026-128PK 18.45 USD
470026-128 470026-132 470026-130
Ward's® Serpentinite
Specimens Rock Specimens Metamorphic Rocks
Dark-green “verde antique”, containing some chlorite and calcite.
Ordering information: WARD’S offers different specimen sizes to meet your various needs. Student specimens are ideal for students to use for study and identification. Student specimens range in size from 1×1" to 1×2" and come in packages of ten. Hand specimens are great for lectures, demonstrations, and display. Hand specimens range in size from 2×3" to 3×4", and each comes in a labeled plastic tray. Bulk pack includes several of the smaller pieces that are suitable for classroom study, while the larger specimens in the one kilogram pack can be cut to sizes that meet your specific needs. The number and size of pieces will vary.
Caution: Due to the nature of this product it may contain asbestos.