Ward's® Rotary Lapidary Tumbler, 6 lbs. Capacity
Supplier: LORTONE, INC.
006-090
470022-236EA 336.4 USD
470022-236
Ward's® Rotary Lapidary Tumbler, 6 lbs. Capacity
Educational Materials Earth Science Educational Materials Minerals and Rocks Learning Activities
Compact, Portable, and Economical
Its self-contained motor, light weight, and quiet all-rubber barrel let you operate this compact machine virtually anywhere. The generous 6 lb. capacity with simple snap-off lid cuts grit changing time. With 1/20 hp motor, belt cover, precision barrel guide, and cushion feet. 120 VAC, with a 6' cord; 60 Hz. Size: 11 1/2"L x 8"W x 10 1/2"H; weight: 12 lbs.