Ward's® Plant Press
506.4000.1 5267908 B
470001-816EA 79.2 USD
470001-816 470105-318
Ward's® Plant Press
Presses
Professional Quality For Botanical Collecting
This classic botanist’s aid adheres to a key of effective specimen preparation: quick drying ensures a better end result. In our model, special blotter sheets are extra absorbent for faster drying and better color retention. Ventilators provide even airflow around specimens, which also speeds the drying process, while at the same time preventing shrinkage. The large size allows you to preserve complete specimens, while the reinforced 1 wooden frame and cloth straps apply even pressure for quality results.
The press will hold thirty sheets and comes with the hardwood frame, two wear-resistant heavy cloth straps, twelve blotter sheets, and twelve corrugated ventilator sheets