Ward's® Live Plant Kingdom Review Set
Supplier: VWR
WARD470176-292
470176-292KT 125.95 USD
470176-292
Ward's® Live Plant Kingdom Review Set
Specimens Live Specimens Plants - Living Material
Contains Representatives from All Major Plant Groups
Compare the different types of specimens within the plant kingdom with the wide selection included in this set. Keep them in the pots supplied or plant them in a terrarium.
The set of 15 includes dicots (2), monocots (2), insectivorous plant, lichen, liverwort, mangrove propagules (3), Selaginella, moss, fern, Equisetum, and Volvox culture, as well as care instructions.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Check with your state’s Department of Agriculture for any state restrictions.
Delivery information: Please specify a delivery date during checkout.
This item not available in Canada