Multiplate Hester-Dendy Sampler
244283
470021-286EA 75.95 USD
470021-286
Multiplate Hester-Dendy Sampler
Approved by the US Geological Survey and EPA for Benthic Macroinvertebrate Collection
Plates and samplers in both designs are made of 1/8" smooth, tempered hardboard. The eyebolt, washers, and wingnut are made of stainless steel for long life. Wingnuts at the bottom allow disassembly without tools.