Motorized, Illuminated Planetarium
Supplier: SCIENCE INTERACTIVE GROUP
653-3020
470006-098EA 1231 USD
470006-098
Motorized, Illuminated Planetarium
Models Astronomy Models
Completely Automated Instrument with Synchronized Orbits
  • Model has a built in compass
  • Sun provides illumination
Demonstrate the motions of the Earth, moon, sun, and Venus. The model features synchronized orbital movements for the sun, Earth, and Venus; illuminated 6"-diameter sun; and a built-in compass. 115 VAC with a 5' cord. Length: 16 1/2".