Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Geo-logic Metamorphic Rock Set

Supplier: VWR
WARD470175-722 WARD470025-352
Altered Igneous and Sedimentary Rocks are Clearly Represented

  • Developed jointly by experienced educators and Ward's® staff geologists
  • Classroom tested for dependability
  • Designed to save valuable planning and preparation time

The GEO-logic activities in this set will help students learn to identify different types of metamorphic rocks by studying their features and composition with our GEO-logic collection. A teacher's guide and student copymasters are included for the following activities: determining causes of foliation, classification and identification by texture and composition, and deciphering rock history. The set is housed in a compartmented collection box.

GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides

The GEO-logic system provides a modular approach to teaching and allows you to address broad curriculum needs for the study of minerals and rocks; natural resources and energy; and fossils and earth history. The two types of GEO-logic teaching collections include:
• A Reference Set, which provides an overview of all topics and comes with a teacher’s guide
• Topic Sets, which provide greater depth in an individual subject area and include both a teacher’s guide and student copymasters
• Classroom Bundles, which provide 15 bulk-packaged samples included in the reference or topic set and 15 sets of any required testing materials included with the corresponding set
*Classroom Bundles are bulk-packaged and do not include display boxes, or teacher/student activity guides.

Contents:
12 individually numbered samples, 1 x 2"
Rocks:amphibolite, coal (anthracite), gneiss (3), marble, quartzite, schist (2), serpentinite, slate, soapstone
Magnifier"



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Metamorphic Rocks Topic Set (12 Specimens)   470025-352 Each Retrieving Restricted
Metamorphic Rocks Classroom Bundle (15 of 12 Unique Specimens)   470175-722 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Size 1 x 2"

ACCESSORIES

Expand All / Collapse All

Accessories
Description Dimensions No. of Slides Supplier No. VWR Catalog Number Unit Price Quantity
Metamorphic Rocks Thin Sections 27 x 46 mm 12   470025-358 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty