Marine Salt Mix
Supplier: VWR
WARD470154-008
470154-008PK 6.95 USD
470154-008
Marine Salt Mix
Specimens Live Specimens Aquaria - Living Material
Create a suitable home for fish, brine shrimp and other marine invertebrates. A 450 g bag will make three gallons of sea water (1.020-1.025 specific gravity). (Please note: This item is recommended for use with saltwater aquaria only.)