Magnifier 5x Illuminated LED
1185-50
470128-280EA 32.65 USD
470128-280
Magnifier 5x Illuminated LED
Magnifiers
This multiuse magnifier is perfect for everyday home or office use to read fine print and see details. It slides over the paper so you won’t need to hold the handle, and has 4 LED lights providing an illuminated viewing area. With transparent sides to allow light in, it can also be used for examining specimens in the science classroom and biology laboratory.
  • 5x magnification
  • Great for travel
  • Easy-to-use
The magnifier has strong acrylic construction and has a 5x glass lens that is 3" (75 mm) in diameter. Comes with a semi-hard zipped case for secure storage. Product is not a toy. It is for laboratory/educational use only. It is not for use by children 12 and under. Requirements: 1 CR2032 button cell battery, included