Lipase

Supplier: ALDON CORP SE
LL0200-25G
470301-376EA 20.05 USD
470301-376
Lipase
Chemicals for Science Education
CAS Number: 9001-62-1
Solubility: Water
Synonyms: Lipase Enzyme
Shelf Life (months): 12
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR