Would you like to log in again?
Laser Pointer
Supplier: ONPOINT LASERS, INC.
SRLP-B
470096-998EA 20.85 USD InStock
470096-998
Laser Pointer
Laser Pointers
This stylish Class IIIa laser pointer has a great looking executive design with black body and gold trim. Minimum output is <5mW. Wavelength: 630-680nm. Features on/off button and pocket clip. Comes in plastic snap-case with 2 AAA batteries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty