Illuminated Colony Counter
10
470200-452EA 184.95 USD
470200-452
Illuminated Colony Counter
Colony Counters
The translucent viewing screen is imprinted with standard Wolfhuegel rulings that form 1 cm squares with diagonal rows subdivided into 9 equal squares.
Will accommodate petri dishes up to 125mm in diameter. 110V AC.
Delivery information: Includes: lamp, line cord with on/off switch.