Flower Structure Riker Mount, Ward’s®
Supplier: Riker Mount
R155
470101-644EA 164.8 USD
470101-644
Flower Structure Riker Mount, Ward’s®
Specimens Preserved Specimens Preserved Plant Materials
Easy review of floral structure
  • Easy to store
  • Real dissected specimen
This attractive, expertly mounted display features a tulip, carefully dissected to show the flowers structure, male parts, female parts and internal structure. A labeled diagram of a whole tulip identifies each part and shows how it fits in the whole.
Ordering information: As these are prepared from real specimens, the product you receive may appear different from the photograph.