Fiber Identification Kit
WARD470104-662
470104-662KT 187.99 USD
470104-662
Fiber Identification Kit
Educational Classroom Kits and Activities
Feel like a real forensic professional while using this kit.
  • Grades 7-12
  • Examine physical and chemical properties
  • Estimated class/lab time required is 120 minutes
Turn your classroom into a forensics lab with this fiber analysis kit. Aspiring forensic scientists will perform qualitative tests on six different materials to sharpen their knowledge of physical and chemical properties, soluability and more. In addition to emphasis on applying chemistry principles to forensics, this lab activity also helps develop proper laboratory technique.
The Fiber Identificatioin Kits was created by Teacher Developer Tim Graham. Kit includes enough materials for 4 classes of 10 groups.
Materials included:
(6) fiber samples
Dupont #4 Fiber Identification Stain
Calcium hydroxide
Benedict's solution
Sodium hydroxide
Lead acetate
Copper sulfate
Formic acid
Acetone
Litmus paper
Instructions
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.