EdCam Digital Gel Imaging Systems
Supplier: PECA PRODUCTS INC.
EDCAM-8 EDCAM-7 EDCAM 10
470015-074EA 952.89 USD
470015-074 470015-072 470012-910
EdCam Digital Gel Imaging Systems
Photodocumentation Systems
Digitally photograph your gels for a permanent record of results.
  • High quality digital cameras and hoods
  • Transilluminators not included
Each EdCam system includes a current digital camera and a hood specifically designed for this system with a built in close-up lens that assures proper focus for a sharp clear image of the gel. System 8 hood has a specially engineered filter that blocks unwanted UV and Infrared light, necessary to assure bands in the gel can be seen. The hood blocks outside light from reflecting off the gels surface so room lights can remain on during use. Hood will accommodate gels up to 7.9×9.8cm (31/8×37/8").