Desiccant Cartridges
Supplier: Bel-Art Products, a part of SP
F42048-0065
470121-958PK 90.05 USD
470121-958
Desiccant Cartridges
Desiccators Desiccants
Contain Silica Gel in a Clear Plastic Container.
  • Cartridges are 8-mesh
  • Regenerate cartridges by heating or can be discarded
A blue to pink color change in silica gel indicates the presence of moisture and need for changing. Pkg. of 6.