Would you like to log in again?
Ward's® Deluxe Berlese Apparatus
New, More Efficient Design
Extract insects and small invertebrates from soil samples or leaf litter most effectively. Using this deluxe model, you can place a larger amount of matrix in the apparatus at one time; a plastic screen is enclosed by durable muslin material to keep the matrix trapped while the apparatus is in use. Simply dump the matrix through the opening in the top of the apparatus and pull the draw string closed. As you grip the two 8" handles on opposite sides of the apparatus and shake, organisms will fall through the screen, down a fine polyester netting, and land in a receptical you provide secured at the tool’s other end with the integral draw string. The unit is collapsible for easy storage and transport. Overall size: 12 1/4"dia. x 40"L.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
WARD’S Deluxe Berlese Apparatus P24317 470018-454 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty