Bial’s Reagent

Supplier: ALDON CORP SE
BB0115-100ML
470300-378EA 22.65 USD
470300-378
Bial’s Reagent
Chemicals for Science Education
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Flammable, Corrosive
Synonyms: Bial's Test Reagent
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR