Soldering Iron 30 W
400-110
470123-386EA 32.6 USD
470123-386
Soldering Iron 30 W
Soldering Supplies Soldering Irons
For intricate soldering; 10" long with nickel plated cone tip WLS-75100-26A. Has 6' long 2-wire cord with plug. For 115 V, 50/60 Hz. UL listed.