Paraffin wax, medium flakes
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Wax Paraffin
, Paraffin wax
W1061-1LB
700000-500EA
77
USD
700000-500
Paraffin wax, medium flakes
Paraffin
Wax Paraffin, Pastilles consists of a mix of hydrocarbon molecules and is derived from coal, petroleum, or oil shale. It can be used in electrical insulation, lubrication and candles.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00132833 CAS Number: 8002-74-2 EINECS: 232-315-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...