Paraffin wax, white, cake
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Wax Paraffin
, Paraffin wax
W1025-1LB
700000-496EA
89
USD
700000-496
Paraffin wax, white, cake
Paraffin
Wax Paraffin, White, Cake consists of a mix of hydrocarbon molecules and is derivable from coal, petroleum, or oil shale. It is used in electrical insulation, lubrication and candles.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
0.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00132833 CAS Number: 8002-74-2 EINECS: 232-315-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...