DL-α-Tocopherol acetate FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
, DL-alpha-Tocopheryl acetate
, Vitamine E acetate
All-rac-a-Tocopheryl Acetate, FCC is the synthetic form of Vitamin E. Alpha-tocopheryl acetate is more resistant to oxidation during storage than unesterified tocopherols. The bioavailability of alpha-tocopherol from alpha-tocopheryl acetate is roughly equivalent to that of free alpha-tocopherol. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₃₁H₅₂O₃ MW: 472.75 g/mol Density: 0.96 Storage Temperature: Ambient |
MDL Number:
MFCD00072052 CAS Number: 7695-91-2 EINECS: 231-710-0 |
Specification Test Results
Assay | 96% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...