(±)-α-Terpineol ≥85%, Practical grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 2-(4-Methylcyclohex-3-en-1-yl)propan-2-olTerpineol

TE107-4LTGL TE107-500MLG TE107-55002 TE107-200LTB TE107-20LTMT
700016-050EA 176 USD
700016-050 700016-046 700016-044 89092-200 700016-048
(±)-α-Terpineol ≥85%, Practical grade
(±)-α-Terpineol

alpha-Terpineol, Practical, is a naturally occurring monoterpene alcohol often isolated from pine oil. The Alpha isomer is one of three isomers, and is generally the most common constituent. It has a pleasenat lilac odor and for this reason is often used in perfumes, cosmetics and flavoring agents

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Density: 0,933
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6

Order Now

Specification Test Results

Assay 85% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR