Triclosan ≥97% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 5-Chloro-2-(2, 4-dichlorophenoxy) phenol5-Chloro-2-(2,4-dichlorophenoxy)phenol

T1589-25KGBL
700003-628EA 9933 USD
700003-628
Triclosan ≥97% USP
Triclosan

Triclosan, USP is used as an antibacterial and antifungal agent, as well as a selective agent in molecular cloning. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₇Cl₃O₂
MW: 289.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800992
CAS Number: 3380-34-5
EINECS: 222-182-2

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR