Triethylene glycol monomethyl ether ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: mPEG3-AlcoholMethyl triglycol2-(2-(2-Methoxyethoxy)ethoxy)ethanol

T1335-2LT T1335-100ML T1335-500ML
700003-570EA 126 USD
700003-570 700003-566 700003-568
Triethylene glycol monomethyl ether ≥95%
Triethylene glycol monomethyl ether

Triethylene Glycol Methyl Ether, also known as TEG or triglycol, is a viscous liquid used in air sanitizer products and as a plasticizer for vinyl, Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₇H₁₆O₄
MW: 164.2 g/mol
Density: 1,026
Storage Temperature: Ambient
MDL Number: MFCD00002875
CAS Number: 112-35-6
EINECS: 203-962-1

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR