Sodium chlorate ≥99%, crystals, Reagent Grade

Danger

S1235-12KG S1235-500GM S1235-2.5KG S1235-45KGBL S1235-125GM
700001-354EA 258.96 USD
700001-354 700001-352 700001-350 700001-348 700001-346
Sodium chlorate ≥99%, crystals, Reagent Grade
Sodium chlorate

Form: Crystal

Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 260 °C
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1,II
REACH: 01-2119474389-23
Merck Index: 12,08741

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR