Retinol all trans ≥95%

Synonyms: (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-olall-trans-RetinolVitamin A alcohol

R1474-100MG R1474-500MG R1474-1GM
700009-320EA 379.74 USD
700009-320 700009-322 700009-318
Retinol all trans ≥95%
Retinol all trans

all-trans-Retinol is also known as all-trans-retinol 13,14-reductase. This is an enzyme that belongs to the family of oxidoreductases. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₂₀H₃₀O
MW: 286.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00001552
CAS Number: 68-26-8
EINECS: 200-683-7

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR