Perfluorodecahydronaphthalene (mixture of cis and trans isomers) ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Perfluoro(decahydronaphthalene)PerfluorodecahydronaphthalenePerfluorodecalin

P2310-25GM P2310-100GM
700002-460EA 95 USD
700002-460 700002-458
Perfluorodecahydronaphthalene (mixture of cis and trans isomers) ≥95%
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)

Perfluorodecalin, also known as Flutec PP6 or F-Decalin, is a fluorocarbon and a derivative of decalin which is most often used to dissolve gases. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₀F₁₈
MW: 462.08 g/mol
Density: 1.908
Storage Temperature: Ambient
MDL Number: MFCD00010626
CAS Number: 306-94-5
EINECS: 206-192-4

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR