D-Panthenol ≥98% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Dexpanthenol
, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
P1831-100GM
P1831-1KG
95032-852EA
1017
USD
95032-852
95032-850
D-Panthenol ≥98% USP
D-Panthenol
Dexpanthenol, USP is the alcohol analog of pantothenic acid, and is thus a provitamin of B5.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₉H₁₉NO₄ MW: 205.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065006 CAS Number: 81-13-0 EINECS: 201-327-3 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...