Pyridine-2-aldoxime methochloride ≥97% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-PAM chloride2-((Hydroxyimino)methyl)-1-methylpyridin-1-ium chloridePralidoxime ChloridePyridine-2-carboxaldoxime methochloride

P1722-5GM P1722-1KGBL P1722-500GM P1722-1GM P1722-25GM P1722-100GM
95035-268EA 1507 USD
95035-268 95035-264 95035-266 95035-270 95035-262 75812-000
Pyridine-2-aldoxime methochloride ≥97% USP
Pyridine-2-aldoxime methochloride

Pralidoxime Chloride, USP is used as an antidote to treat poisoning by a chemical or pesticide insect spray.


All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011981
CAS Number: 14018-50-9
EINECS: 200-080-9

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR