Potassium citrate monohydrate 99.0-100.5%, granular FCC

Synonyms: Tripotassium citrate monohydratePotassium citrate tribasic monohydrateCitric acid tripotassium salt monohydrateTripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate

P1266-500GM P1266-2.5KG
95035-146EA 554 USD
95035-146 95035-148
Potassium citrate monohydrate 99.0-100.5%, granular FCC
Potassium citrate monohydrate

Potassium Citrate, Monohydrate, Granular, FCCis used in foods and beverages as buffering, sequestering or emulsifying agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOC(COOK)(CH₂COOK)₂·H₂O
MW: 324.41 g/mol
Melting Pt: 230 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
Merck Index: 13,07706
https://us.vwr.com/stibo/web/std.lang.all/72/37/2297237.jpg

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR