2-Ethylhexyl salicylate ≥95% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Ethylhexyl 2-hydroxybenzoateSalicylic acid Octyl esterSalicylic acid 2-Ethylhexyl esterOctyl Salicylate

O1236-500ML O1236-4LT O1236-100ML O1236-500ML O1236-4LT O1236-20KGBL O1236-20KGBL O1236-100ML
95034-772EA 291 USD
CA95034-772 95034-772 CA95034-770 95034-770 700006-428 CA700006-428 CA95034-768 95034-768
2-Ethylhexyl salicylate ≥95% USP
2-Ethylhexyl salicylate

Octisalate, USP absorbs ultraviolet radiation in the UVB range and is used in sunscreens and other cosmetics to protect from skin damage. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053300
CAS Number: 118-60-5
EINECS: 204-263-4

Order Now

Specification Test Results

Assay 95% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR