Ethanolamine hydrochloride ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Aminoethanol hydrochloride
, 2-Hydroxyethylammonium Chloride
, Monoethanolamine Hydrochloride
M1456-100GM
M1456-25GM
700006-922EA
83
USD
700006-922
700006-920
Ethanolamine hydrochloride ≥98%
Ethanolamine hydrochloride
Monoethanolamine Hydrochloride, also known as ethanolamine or 2-aminoethanol, is an organic chemical compound used as feedstock in the production of emulsifiers, detergents, and pharmaceuticals.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₂H₈ClNO MW: 97.54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012892 CAS Number: 2002-24-6 EINECS: 217-900-6 |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...