Ketoprofen ≥98.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-(3-Benzoylphenyl)propanoic acid2-(3-Benzoylphenyl)propionic acid

K1155-100GMP K1155-25KGBL K1155-1GMGL K1155-25GMPL
95031-144EA 597.08 USD
95031-144 89078-826 95031-140 95031-138
Ketoprofen ≥98.5% USP
Ketoprofen

Ketoprofen, USP is a nonsteroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055790
CAS Number: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR