α-Ionone

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

I2471-100GM I2471-25GM
700003-966EA 120.35 USD
700003-966 700003-968
α-Ionone
α-Ionone

alpha-Ionone is used as a fragrance in perfumes, cosmetics and personal care products, as well as in household cleaners and detergents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₃H₂₀O
MW: 192.3 g/mol
Density: 0.931
Storage Temperature: Ambient
MDL Number: MFCD00001565
CAS Number: 127-41-3
EINECS: 204-841-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR